Assay Details
Target Gene Details
Entrez Gene ID: | 54511 |
Gene Name: | 3-hydroxymethyl-3-methylglutaryl-CoA lyase like 1 |
Gene Aliases: |
bA418P12.1, er-cHL |
Location: |
Chr.6:55434373-55579214 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HMGCLL1 | NM_001042406.1 | NP_001035865.1 | ||
NM_001287741.1 | NP_001274670.1 | |||
NM_001287746.1 | NP_001274675.1 | |||
NM_001287753.1 | NP_001274682.1 | |||
NM_019036.2 | NP_061909.2 | |||
NR_109869.1 | ||||
AF131827.1 | AAD20054.1 | |||
AK055075.1 | ||||
AK127587.1 | BAC87045.1 | |||
AK293856.1 | ||||
AK294210.1 | ||||
AK297191.1 | ||||
BC024194.2 | AAH24194.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1018622 | Chr.6:55433056 - 55455513 on Build GRCh38 | Loss | HMGCLL1 |
dgv5965n100 | Chr.6:55372925 - 55522683 on Build GRCh38 | Gain | HMGCLL1 GFRAL |
nsv470819 | Chr.6:55372139 - 55504017 on Build GRCh38 | Gain | HMGCLL1 GFRAL |
nsv1034037 | Chr.6:55357611 - 56204802 on Build GRCh38 | Gain | COL21A1 HMGCLL1 BMP5 GFRAL |
nsv462948 | Chr.6:55373293 - 55504017 on Build GRCh38 | Gain | HMGCLL1 GFRAL |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |