Assay Details
Target Gene Details
Entrez Gene ID: | 100996485 |
Gene Name: | chromosome 5 open reading frame 66 |
Gene Aliases: |
- |
Location: |
Chr.5:135033280-135344680 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
C5orf66 | NM_001277348.1 | NP_001264277.1 | ||
AK026965.1 | BAB15606.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv823239 | Chr.5:135019535 - 135071110 on Build GRCh38 | Gain | C5orf66-AS1 C5orf66 PITX1 |
nsv830495 | Chr.5:135024595 - 135204207 on Build GRCh38 | Loss | C5orf66-AS1 C5orf66 PITX1 |
dgv969n67 | Chr.5:135018061 - 135059172 on Build GRCh38 | Gain | C5orf66-AS1 C5orf66 PITX1 |
nsv599744 | Chr.5:135012266 - 135070292 on Build GRCh38 | Loss | C5orf66-AS1 C5orf66 PITX1 |