Assay Details
Target Gene Details
Entrez Gene ID: | 6932 |
Gene Name: | transcription factor 7 (T-cell specific, HMG-box) |
Gene Aliases: |
TCF-1 |
Location: |
Chr.5:134114703-134148229 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv599732 | Chr.5:134099223 - 134230401 on Build GRCh38 | Loss | MIR3661 SKP1 TCF7 PPP2CA |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |