Assay Details
Target Gene Details
Entrez Gene ID: | 839 |
Gene Name: | caspase 6 |
Gene Aliases: |
MCH2 |
Location: |
Chr.4:109688629-109703473 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CASP6 | NM_001226.3 | NP_001217.2 | ||
NM_032992.2 | NP_116787.1 | |||
NR_133012.1 | ||||
BC000305.2 | AAH00305.1 | |||
BC004460.1 | AAH04460.1 | |||
BM837197.1 | ||||
BX380795.2 | ||||
U20536.1 | AAC50168.1 | |||
U20537.1 | AAC50169.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv595142 | Chr.4:109688325 - 109735574 on Build GRCh38 | Loss | CASP6 PLA2G12A MCUB CFI |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |