Assay Details
Target Gene Details
Entrez Gene ID: | 86 |
Gene Name: | actin like 6A |
Gene Aliases: |
ACTL6, ARPN-BETA, Arp4, BAF53A, INO80K |
Location: |
Chr.3:179562880-179588405 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ACTL6A | NM_004301.4 | NP_004292.1 | ||
NM_177989.3 | NP_817126.1 | |||
NM_178042.3 | NP_829888.1 | |||
AA256726.1 | ||||
AB015907.1 | BAA74577.1 | |||
AB060168.1 | BAB87844.1 | |||
AB061315.1 | BAB87848.1 | |||
AF041474.1 | AAC94991.1 | |||
AK021756.1 | ||||
AK098691.1 | ||||
AK223212.1 | BAD96932.1 | |||
AL136608.1 | CAB66543.1 | |||
BC000949.2 | AAH00949.1 | |||
BC001391.2 | AAH01391.1 | |||
BC036371.1 | AAH36371.1 | |||
CR533529.1 | CAG38560.1 | |||
DB041143.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |