Assay Details
Target Gene Details
Entrez Gene ID: | 22906 |
Gene Name: | trafficking kinesin protein 1 |
Gene Aliases: |
MILT1, OIP106 |
Location: |
Chr.3:42013802-42225890 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TRAK1 | XM_017005906.1 | XP_016861395.1 | ||
XM_017005909.1 | XP_016861398.1 | |||
XM_017005911.1 | XP_016861400.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv460520 | Chr.3:41918514 - 42139904 on Build GRCh38 | Gain | TRAK1 ULK4 |
esv2760776 | Chr.3:41872696 - 42174077 on Build GRCh38 | Gain | TRAK1 ULK4 |
dgv8295n54 | Chr.3:41843258 - 42142869 on Build GRCh38 | Gain | TRAK1 ATP6V0E1P2 ULK4 |
esv3595984 | Chr.3:41873454 - 42129694 on Build GRCh38 | Gain | TRAK1 ULK4 |
dgv8296n54 | Chr.3:41859164 - 42073996 on Build GRCh38 | Gain | TRAK1 ATP6V0E1P2 ULK4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |