Assay Details
Target Gene Details
Entrez Gene ID: | 55179 |
Gene Name: | Fas apoptotic inhibitory molecule |
Gene Aliases: |
FAIM1 |
Location: |
Chr.3:138608163-138633376 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FAIM | NM_001033030.1 | NP_001028202.1 | ||
NM_001033031.1 | NP_001028203.1 | |||
NM_001033032.1 | NP_001028204.1 | |||
NM_018147.3 | NP_060617.1 | |||
XM_011512950.2 | XP_011511252.1 | |||
AK001444.1 | BAA91695.1 | |||
AK125477.1 | 5 | 1951 | BAC86174.1 | |
BC012478.1 | AAH12478.1 | |||
BM738750.1 | ||||
BQ638715.1 | ||||
BU738945.1 | ||||
CR457122.1 | CAG33403.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv591854 | Chr.3:138629239 - 139045387 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv428422 | Chr.3:137344347 - 138686784 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv3597916 | Chr.3:138478438 - 138638584 on Build GRCh38 | Gain |
![]() ![]() ![]() |
dgv4908n100 | Chr.3:138494324 - 139018846 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)