Assay Details
Target Gene Details
Entrez Gene ID: | 112616 |
Gene Name: | CKLF like MARVEL transmembrane domain containing 7 |
Gene Aliases: |
CKLFSF7 |
Location: |
Chr.3:32391671-32455528 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CMTM7 | NM_138410.3 | NP_612419.1 | ||
NM_181472.2 | NP_852137.1 | |||
XM_011533319.2 | XP_011531621.1 | |||
XM_017005646.1 | XP_016861135.1 | |||
AF479263.2 | AAN73041.1 | |||
AK308878.1 | ||||
AY174113.1 | AAO44955.1 | |||
BC010116.2 | AAH10116.1 | |||
DQ132881.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv518025 | Chr.3:32398293 - 32415706 on Build GRCh38 | Gain | CMTM7 |
nsv1003285 | Chr.3:31386933 - 33172886 on Build GRCh38 | Gain | OSBPL10-AS1 CMTM6 CCR4 CNOT10 TRIM71 GPD1L LOC105377022 STT3B OSBPL10 GLB1 CMTM8 CRTAP LOC105377021 TMPPE SUSD5 ZNF860 CMTM7 MIR548AY DYNC1LI1 |
nsv954461 | Chr.3:32396709 - 32428608 on Build GRCh38 | Duplication | CMTM7 |
nsv834652 | Chr.3:32375792 - 32564922 on Build GRCh38 | Loss | CMTM6 CMTM7 MIR548AY DYNC1LI1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |