Assay Details
Target Gene Details
Entrez Gene ID: | 339926 |
Gene Name: | EHHADH antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:185162901-185191955 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EHHADH-AS1 | NR_038990.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv5002n100 | Chr.3:185129229 - 185287036 on Build GRCh38 | Gain | EHHADH-AS1 MIR5588 C3orf70 MAP3K13 EHHADH |
esv3598965 | Chr.3:185185472 - 185197830 on Build GRCh38 | Gain | EHHADH-AS1 EHHADH |