Assay Details
Target Gene Details
Entrez Gene ID: | 55802 |
Gene Name: | decapping mRNA 1A |
Gene Aliases: |
HSA275986, Nbla00360, SMAD4IP1, SMIF |
Location: |
Chr.3:53283423-53347627 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DCP1A | NM_001290204.1 | NP_001277133.1 | ||
NM_001290205.1 | NP_001277134.1 | |||
NM_001290206.1 | NP_001277135.1 | |||
NM_001290207.1 | NP_001277136.1 | |||
NM_018403.6 | NP_060873.4 | |||
AA129009.1 | ||||
AB075507.1 | BAE45754.1 | |||
AJ275986.1 | CAB77023.1 | |||
AK001969.1 | BAA92008.1 | |||
AK295205.1 | ||||
AY146651.1 | AAN62763.1 | |||
BC007439.2 | AAH07439.1 | |||
BI769171.1 | ||||
BP284272.1 | ||||
KU178716.1 | ||||
KU178717.1 | ||||
KU178718.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |