Assay Details
Target Gene Details
Entrez Gene ID: | 23228 |
Gene Name: | phospholipase C like 2 |
Gene Aliases: |
PLCE2 |
Location: |
Chr.3:16802691-17090606 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PLCL2 | XM_017006023.1 | XP_016861512.1 | ||
AK309946.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)