Assay Details
Target Gene Details
Entrez Gene ID: | 92106 |
Gene Name: | oxidoreductase NAD binding domain containing 1 |
Gene Aliases: |
- |
Location: |
Chr.3:16265160-16386979 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
OXNAD1 | XM_011534232.2 | XP_011532534.1 | ||
XM_011534233.2 | XP_011532535.1 | |||
XM_011534234.2 | XP_011532536.1 | |||
XM_017007485.1 | XP_016862974.1 | |||
XM_017007488.1 | XP_016862977.1 | |||
XM_017007489.1 | XP_016862978.1 | |||
XM_017007491.1 | XP_016862980.1 | |||
XM_017007494.1 | XP_016862983.1 | |||
XM_017007496.1 | XP_016862985.1 | |||
AK298829.1 | ||||
DQ364598.1 | ABC95193.1 |
Target Gene Details
Entrez Gene ID: | 23180 |
Gene Name: | raftlin, lipid raft linker 1 |
Gene Aliases: |
MIG2, PIB10, PIG9, RAFTLIN |
Location: |
Chr.3:16315845-16513715 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RFTN1 | NM_015150.1 | NP_055965.1 | ||
XM_005264985.1 | XP_005265042.1 | |||
XM_005264986.2 | XP_005265043.1 | |||
XM_006713069.1 | XP_006713132.1 | |||
XM_011533529.1 | XP_011531831.1 | |||
XM_011533530.1 | XP_011531832.1 | |||
XM_011533531.1 | XP_011531833.1 | |||
XM_017006005.1 | XP_016861494.1 | |||
XM_017006006.1 | XP_016861495.1 | |||
XM_017006007.1 | XP_016861496.1 | |||
AK074446.1 | ||||
AK301024.1 | ||||
AK303846.1 | ||||
AK310258.1 | ||||
AY237654.1 | AAO91814.1 | |||
BC006400.2 | AAH06400.4 | |||
BC014032.2 | AAH14032.2 | |||
BC032349.1 | AAH32349.1 | |||
BC051336.1 | AAH51336.1 | |||
BC069209.1 | AAH69209.1 | |||
BC098270.1 | ||||
BC100667.1 | ||||
BC100669.1 | ||||
BC121795.1 | ||||
D42043.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834623 | Chr.3:16313913 - 16474057 on Build GRCh38 | Gain | RFTN1 OXNAD1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |