Assay Details
Target Gene Details
Entrez Gene ID: | 2047 |
Gene Name: | EPH receptor B1 |
Gene Aliases: |
ELK, EPHT2, Hek6, NET |
Location: |
Chr.3:134776089-135260467 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EPHB1 | XM_017005868.1 | XP_016861357.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)