Assay Details
Target Gene Details
Entrez Gene ID: | 10225 |
Gene Name: | CD96 molecule |
Gene Aliases: |
TACTILE |
Location: |
Chr.3:111542079-111665996 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CD96 | NM_001318889.1 | NP_001305818.1 | ||
NM_005816.4 | NP_005807.1 | |||
NM_198196.2 | NP_937839.1 | |||
NR_134917.1 | ||||
XM_005247063.3 | XP_005247120.1 | |||
XM_006713469.3 | XP_006713532.1 | |||
XM_006713470.3 | XP_006713533.1 | |||
XM_017005521.1 | XP_016861010.1 | |||
XM_017005522.1 | XP_016861011.1 | |||
AK290719.1 | ||||
AK315237.1 | ||||
AL833681.1 | CAI46155.1 | |||
BC020749.1 | AAH20749.1 | |||
BM561433.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv460821 | Chr.3:110918624 - 111782250 on Build GRCh38 | Gain | ZBED2 NECTIN3 LOC151760 CD96 PHLDB2 PLCXD2 PLCXD2-AS1 PVRL3-AS1 |
nsv3946 | Chr.3:111517188 - 111564191 on Build GRCh38 | Deletion | CD96 |
nsv997756 | Chr.3:111507211 - 111657738 on Build GRCh38 | Gain | ZBED2 CD96 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |