Assay Details
Target Gene Details
Entrez Gene ID: | 30849 |
Gene Name: | phosphoinositide-3-kinase regulatory subunit 4 |
Gene Aliases: |
VPS15, p150 |
Location: |
Chr.3:130678934-130746852 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PIK3R4 | NM_014602.2 | NP_055417.1 | ||
AK314219.1 | ||||
BC110318.1 | AAI10319.1 | |||
BC127105.1 | ||||
BC127106.1 | ||||
Y08991.1 | CAA70176.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv34166 | Chr.3:130374350 - 130769777 on Build GRCh38 | Loss | COL6A5 PIK3R4 COL6A6 |
nsv460855 | Chr.3:130627735 - 130748727 on Build GRCh38 | Loss | PIK3R4 COL6A6 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |