Assay Details
Target Gene Details
Entrez Gene ID: | 2632 |
Gene Name: | glucan (1,4-alpha-), branching enzyme 1 |
Gene Aliases: |
APBD, GBE, GSD4 |
Location: |
Chr.3:81489699-81761799 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GBE1 | NM_000158.3 | NP_000149.3 | ||
AB209731.1 | BAD92968.1 | |||
AK125918.1 | ||||
AK297939.1 | ||||
AK300620.1 | ||||
AK312245.1 | ||||
BC012098.1 | AAH12098.1 | |||
BQ632568.1 | ||||
L07956.1 | AAA58642.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |