Assay Details
Target Gene Details
Entrez Gene ID: | 2042 |
Gene Name: | EPH receptor A3 |
Gene Aliases: |
EK4, ETK, ETK1, HEK, HEK4, TYRO4 |
Location: |
Chr.3:89107524-89482134 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EPHA3 | NM_005233.5 | NP_005224.2 | ||
NM_182644.2 | NP_872585.1 | |||
XM_005264715.2 | XP_005264772.1 | |||
XM_005264716.2 | XP_005264773.1 | |||
AF213459.1 | AAG43576.1 | |||
AF213460.1 | AAG43577.1 | |||
AK291411.1 | ||||
BC063282.1 | AAH63282.1 | |||
M83941.1 | AAA58633.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834760 | Chr.3:89036379 - 89250248 on Build GRCh38 | Loss |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)