Assay Details
Target Gene Details
Entrez Gene ID: | 10644 |
Gene Name: | insulin like growth factor 2 mRNA binding protein 2 |
Gene Aliases: |
IMP-2, IMP2, VICKZ2 |
Location: |
Chr.3:185643739-185825051 on Build GRCh38 |
Assay Gene Location: | Within Intron 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
IGF2BP2 | NM_001007225.1 | NP_001007226.1 | ||
NM_001291869.1 | NP_001278798.1 | |||
NM_001291872.1 | NP_001278801.1 | |||
NM_001291873.1 | NP_001278802.1 | |||
NM_001291874.1 | NP_001278803.1 | |||
NM_001291875.1 | NP_001278804.1 | |||
NM_006548.4 | NP_006539.3 | |||
XM_006713477.3 | XP_006713540.1 | |||
XM_006713478.3 | XP_006713541.1 | |||
XM_017005557.1 | XP_016861046.1 | |||
XM_017005558.1 | XP_016861047.1 | |||
XM_017005559.1 | XP_016861048.1 | |||
XM_017005560.1 | XP_016861049.1 | |||
XM_017005562.1 | XP_016861051.1 | |||
AF117107.1 | AAD09827.1 | |||
AK290664.1 | ||||
AK296703.1 | ||||
BC021290.2 | AAH21290.1 | |||
EU408701.1 | ||||
EU408702.1 | ||||
EU408703.1 | ||||
EU408704.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)