Assay Details
Target Gene Details
Entrez Gene ID: | 8891 |
Gene Name: | eukaryotic translation initiation factor 2B subunit gamma |
Gene Aliases: |
EIF-2B, EIF2Bgamma |
Location: |
Chr.1:44850522-44986722 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EIF2B3 | NM_001261418.1 | NP_001248347.1 | ||
NM_020365.4 | NP_065098.1 | |||
XM_017002745.1 | XP_016858234.1 | |||
XM_017002746.1 | XP_016858235.1 | |||
XM_017002747.1 | XP_016858236.1 | |||
AF257077.1 | AAF91351.1 | |||
AK022399.1 | BAB14027.1 | |||
AK024006.1 | BAB14770.1 | |||
AK314668.1 | ||||
BC018728.1 | AAH18728.1 | |||
BQ776018.1 | ||||
CR457285.1 | CAG33566.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)