Assay Details
Target Gene Details
Entrez Gene ID: | 26027 |
Gene Name: | acyl-CoA thioesterase 11 |
Gene Aliases: |
BFIT, STARD14, THEA, THEM1 |
Location: |
Chr.1:54548134-54634744 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ACOT11 | NM_015547.3 | NP_056362.1 | ||
NM_147161.3 | NP_671517.1 | |||
AB014607.1 | ||||
AF416921.1 | AAL40937.1 | |||
AF416922.1 | AAL40938.1 | |||
AK023937.1 | BAB14734.1 | |||
AK091112.1 | ||||
AK223570.1 | BAD97290.1 | |||
BC001517.2 | AAH01517.1 | |||
BC093844.1 | AAH93844.1 | |||
BC093846.1 | AAH93846.1 | |||
DA121227.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv546266 | Chr.1:54404143 - 54887697 on Build GRCh38 | Gain | MROH7-TTC4 FAM151A LEXM TTC4 ACOT11 TTC22 DHCR24 MROH7 PARS2 SSBP3 |
nsv461583 | Chr.1:54559255 - 54584133 on Build GRCh38 | Gain | ACOT11 |
nsv461572 | Chr.1:54518434 - 54604313 on Build GRCh38 | Gain | ACOT11 |
dgv20e214 | Chr.1:54558682 - 54587087 on Build GRCh38 | Gain | ACOT11 |
nsv829904 | Chr.1:54548613 - 54721384 on Build GRCh38 | Loss | MROH7-TTC4 FAM151A TTC4 ACOT11 MROH7 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |