Assay Details
Target Gene Details
Entrez Gene ID: | 26092 |
Gene Name: | torsin 1A interacting protein 1 |
Gene Aliases: |
LAP1, LAP1B |
Location: |
Chr.1:179882042-179920077 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TOR1AIP1 | NM_001267578.1 | NP_001254507.1 | ||
NM_015602.3 | NP_056417.2 | |||
XM_011509403.1 | XP_011507705.1 | |||
XM_011509404.1 | XP_011507706.1 | |||
AK001780.1 | BAA91906.1 | |||
AK021613.1 | BAB13855.1 | |||
AK023204.1 | BAB14461.1 | |||
AK291495.1 | ||||
AK310012.1 | ||||
BC023247.1 | AAH23247.1 | |||
CN360425.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv473n100 | Chr.1:179814787 - 179895575 on Build GRCh38 | Gain | TOR1AIP2 TOR1AIP1 FAM163A |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |