Assay Details
Target Gene Details
Entrez Gene ID: | 23261 |
Gene Name: | calmodulin binding transcription activator 1 |
Gene Aliases: |
CANPMR |
Location: |
Chr.1:6785324-7769706 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1004598 | Chr.1:6645296 - 7178603 on Build GRCh38 | Gain | DNAJC11 LOC100129476 LOC100505887 CAMTA1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |