Assay Details
Target Gene Details
Entrez Gene ID: | 84976 |
Gene Name: | dispatched RND transporter family member 1 |
Gene Aliases: |
DISPA |
Location: |
Chr.1:222814514-223005995 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DISP1 | NM_032890.3 | NP_116279.2 | ||
XM_011510072.2 | XP_011508374.1 | |||
XM_011510073.2 | XP_011508375.1 | |||
XM_011510074.2 | XP_011508376.1 | |||
XM_011510075.2 | XP_011508377.1 | |||
XM_011510077.2 | XP_011508379.1 | |||
XM_017002611.1 | XP_016858100.1 | |||
DA756409.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv442589 | Chr.1:222843983 - 222850262 on Build GRCh38 | Loss | DISP1 |
nsv1160101 | Chr.1:222843064 - 222850427 on Build GRCh38 | Deletion | DISP1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |