Assay Details
Target Gene Details
Entrez Gene ID: | 64645 |
Gene Name: | major facilitator superfamily domain containing 14A |
Gene Aliases: |
HIAT1 |
Location: |
Chr.1:100038078-100083377 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MFSD14A | NM_033055.2 | NP_149044.2 | ||
XM_005271118.1 | XP_005271175.1 | |||
XM_011541957.1 | XP_011540259.1 | |||
XM_017002084.1 | XP_016857573.1 | |||
AF427492.1 | AAL25115.1 | |||
AK057172.1 | BAB71375.1 | |||
AK096669.1 | BAC04836.1 | |||
BC064409.1 | AAH64409.1 | |||
BC107768.1 | AAI07769.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv830804 | Chr.1:100036777 - 100210481 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
nsv546888 | Chr.1:99954311 - 100084889 on Build GRCh38 | Loss |
![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)