Assay Details
Target Gene Details
Entrez Gene ID: | 7075 |
Gene Name: | tyrosine kinase with immunoglobulin like and EGF like domains 1 |
Gene Aliases: |
JTK14, TIE |
Location: |
Chr.1:43300895-43323110 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TIE1 | NM_001253357.1 | NP_001240286.1 | ||
NM_005424.4 | NP_005415.1 | |||
XM_005271163.1 | XP_005271220.1 | |||
XM_006710869.1 | XP_006710932.1 | |||
XM_017002207.1 | XP_016857696.1 | |||
AF217976.1 | AAG17219.1 | |||
AK225560.1 | ||||
AK300399.1 | ||||
AK300623.1 | ||||
AK307639.1 | ||||
BC038239.1 | AAH38239.1 | |||
DB287371.1 | ||||
EU826586.1 | ||||
EU826587.1 | ||||
EU826588.1 | ||||
EU826589.1 | ||||
EU826590.1 | ||||
X60957.1 | CAA43290.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv546128 | Chr.1:43301467 - 43451738 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv829693 | Chr.1:43129457 - 43312977 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)