Assay Details
Target Gene Details
Entrez Gene ID: | 4919 |
Gene Name: | receptor tyrosine kinase like orphan receptor 1 |
Gene Aliases: |
NTRKR1, dJ537F10.1 |
Location: |
Chr.1:63774019-64181498 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ROR1 | NM_001083592.1 | NP_001077061.1 | ||
NM_005012.3 | NP_005003.2 | |||
AK308088.1 | ||||
BC080541.1 | AAH80541.1 | |||
BC128386.1 | ||||
M97675.1 | AAA60275.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |