Assay Details
Target Gene Details
Entrez Gene ID: | 79368 |
Gene Name: | Fc receptor like 2 |
Gene Aliases: |
CD307b, FCRH2, IFGP4, IRTA4, SPAP1, SPAP1A, SPAP1B, SPAP1C |
Location: |
Chr.1:157745733-157777472 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FCRL2 | NM_001159488.1 | NP_001152960.1 | ||
NM_030764.3 | NP_110391.2 | |||
NR_125358.1 | ||||
XM_011509974.2 | XP_011508276.1 | |||
XM_011509975.2 | XP_011508277.1 | |||
XM_011509976.2 | XP_011508278.1 | |||
XM_017002316.1 | XP_016857805.1 | |||
XM_017002317.1 | XP_016857806.1 | |||
XM_017002318.1 | XP_016857807.1 | |||
XM_017002319.1 | XP_016857808.1 | |||
AF319438.1 | AAK01402.1 | |||
AF390037.1 | AAM12152.1 | |||
AF459633.1 | AAL60249.1 | |||
AK301113.1 | ||||
AL833194.1 | ||||
AY043465.1 | AAK91778.1 | |||
AY358130.1 | AAQ88497.1 | |||
BC069185.1 | AAH69185.1 | |||
BC129836.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |