Assay Details
Target Gene Details
Entrez Gene ID: | 4548 |
Gene Name: | 5-methyltetrahydrofolate-homocysteine methyltransferase |
Gene Aliases: |
HMAG, MS, cblG |
Location: |
Chr.1:236794304-236903981 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MTR | NM_000254.2 | NP_000245.2 | ||
NM_001291939.1 | NP_001278868.1 | |||
NM_001291940.1 | NP_001278869.1 | |||
XM_005273141.4 | XP_005273198.1 | |||
XM_011544194.2 | XP_011542496.1 | |||
XM_017001329.1 | XP_016856818.1 | |||
XM_017001330.1 | XP_016856819.1 | |||
AK294270.1 | ||||
AK299904.1 | ||||
AK307390.1 | ||||
BC130616.1 | ||||
BC136440.1 | ||||
BC144095.1 | ||||
BC144097.1 | ||||
U71285.1 | AAC51188.1 | |||
U73338.1 | AAB39704.1 | |||
U75743.1 | AAB58906.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1005877 | Chr.1:235912335 - 237038508 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv5010 | Chr.1:236796351 - 236810727 on Build GRCh38 | Insertion |
![]() |
nsv428335 | Chr.1:236734659 - 236913672 on Build GRCh38 | Gain |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)