Assay Details
Target Gene Details
Entrez Gene ID: | 55160 |
Gene Name: | Rho guanine nucleotide exchange factor 10 like |
Gene Aliases: |
GrinchGEF |
Location: |
Chr.1:17520197-17697875 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ARHGEF10L | XM_006710728.1 | XP_006710791.1 | ||
XM_006710731.1 | XP_006710794.1 | |||
AK129959.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834302 | Chr.1:17531435 - 17713452 on Build GRCh38 | Loss | ARHGEF10L |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |