Assay Details
Target Gene Details
Entrez Gene ID: | 55970 |
Gene Name: | G protein subunit gamma 12 |
Gene Aliases: |
- |
Location: |
Chr.1:67701466-67833779 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GNG12 | NM_018841.5 | NP_061329.3 | ||
XM_017001809.1 | XP_016857298.1 | |||
XM_017001810.1 | XP_016857299.1 | |||
XM_017001811.1 | XP_016857300.1 | |||
AF119663.1 | AAF17220.1 | |||
AF123766.1 | AAP97245.1 | |||
AF365871.1 | AAK53385.1 | |||
AK223000.1 | BAD96720.1 | |||
AK293101.1 | ||||
AL832431.1 | CAH10645.1 | |||
BC005940.1 | AAH05940.1 | |||
BP236225.1 | ||||
BP236699.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |