Assay Details
Target Gene Details
Entrez Gene ID: | 2048 |
Gene Name: | EPH receptor B2 |
Gene Aliases: |
CAPB, DRT, EK5, EPHT3, ERK, Hek5, PCBC, Tyro5 |
Location: |
Chr.1:22710770-22921500 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EPHB2 | NM_001309192.1 | NP_001296121.1 | ||
NM_001309193.1 | NP_001296122.1 | |||
NM_004442.7 | NP_004433.2 | |||
NM_017449.4 | NP_059145.2 | |||
XM_006710442.3 | XP_006710505.1 | |||
AB210018.1 | ||||
AF025304.1 | AAB94602.1 | |||
AK299701.1 | ||||
BC067861.1 | AAH67861.1 | |||
DB282024.1 | ||||
L36643.1 | AAA74244.1 | |||
L41939.1 | AAA99310.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv34161 | Chr.1:22599762 - 22817730 on Build GRCh38 | Loss | C1QB MIR6127 C1QA EPHA8 MIR4684 EPHB2 C1QC |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |