Assay Details
Target Gene Details
Entrez Gene ID: | 400746 |
Gene Name: | non-compact myelin associated protein |
Gene Aliases: |
C1orf130, MP11 |
Location: |
Chr.1:24556076-24609328 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NCMAP | NM_001010980.4 | NP_001010980.1 | ||
XM_011541463.2 | XP_011539765.1 | |||
AK124519.1 | ||||
CB961010.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv545823 | Chr.1:24568506 - 24621457 on Build GRCh38 | Gain | NCMAP |
nsv834458 | Chr.1:24577828 - 24669644 on Build GRCh38 | Loss | NCMAP SRRM1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |