Assay Details
Target Gene Details
Entrez Gene ID: | 56605 |
Gene Name: | endoplasmic reticulum oxidoreductase 1 beta |
Gene Aliases: |
ERO1LB, Ero1beta |
Location: |
Chr.1:236215122-236282039 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ERO1B | NM_019891.3 | NP_063944.3 | ||
AF252538.1 | AAF97547.1 | |||
AK315351.1 | ||||
BC032823.2 | AAH32823.2 | |||
BC044573.1 | AAH44573.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv590n100 | Chr.1:236139422 - 236267713 on Build GRCh38 | Gain | GPR137B ERO1B |
nsv1005877 | Chr.1:235912335 - 237038508 on Build GRCh38 | Gain | EDARADD GPR137B HEATR1 NID1 MTR LGALS8-AS1 ACTN2 MT1HL1 ERO1B LGALS8 |
nsv522991 | Chr.1:236215849 - 236250146 on Build GRCh38 | Loss | ERO1B |
nsv832969 | Chr.1:236111012 - 236311443 on Build GRCh38 | Gain | GPR137B ERO1B |
nsv549394 | Chr.1:235876577 - 236629706 on Build GRCh38 | Gain | EDARADD GPR137B HEATR1 NID1 LGALS8-AS1 LYST ERO1B LGALS8 |
nsv1012436 | Chr.1:235860657 - 236648100 on Build GRCh38 | Loss | EDARADD GPR137B HEATR1 NID1 LGALS8-AS1 LYST ERO1B LGALS8 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |