Assay Details
Target Gene Details
Entrez Gene ID: | 7042 |
Gene Name: | transforming growth factor beta 2 |
Gene Aliases: |
LDS4, TGF-beta2 |
Location: |
Chr.1:218345334-218444619 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TGFB2 | NM_001135599.2 | NP_001129071.1 | ||
NM_003238.3 | NP_003229.1 | |||
AB209842.1 | BAD93079.1 | |||
AK295671.1 | ||||
AK296504.1 | ||||
AK299839.1 | ||||
AK313106.1 | ||||
BC096235.4 | AAH96235.1 | |||
BC099635.1 | AAH99635.1 | |||
M19154.1 | AAA50404.1 | |||
Y00083.1 | CAA68279.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |