Assay Details
Target Gene Details
Entrez Gene ID: | 481 |
Gene Name: | ATPase Na+/K+ transporting subunit beta 1 |
Gene Aliases: |
ATP1B |
Location: |
Chr.1:169106709-169132722 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ATP1B1 | NM_001677.3 | NP_001668.1 | ||
AK290181.1 | ||||
AK296059.1 | ||||
AK316043.1 | ||||
AY545986.1 | ||||
BC000006.2 | AAH00006.1 | |||
BQ934151.1 | ||||
BT009787.1 | AAP88789.1 | |||
CR407616.1 | CAG28544.1 | |||
U16799.1 | AAC50132.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv22143 | Chr.1:169090390 - 169652030 on Build GRCh38 | Gain+Loss | NME7 BLZF1 F5 SLC19A2 LOC101928596 ATP1B1 CCDC181 SELP |
nsv831858 | Chr.1:169053145 - 169209830 on Build GRCh38 | Gain | LINC00970 NME7 LOC101928596 ATP1B1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |