Assay Details
Target Gene Details
Entrez Gene ID: | 100526835 |
Gene Name: | FPGT-TNNI3K readthrough |
Gene Aliases: |
- |
Location: |
Chr.1:74198212-74544432 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FPGT-TNNI3K | NM_001112808.2 | NP_001106279.2 | ||
NM_001199327.1 | NP_001186256.2 | |||
AB208941.1 | BAD92178.1 | |||
BC032865.1 | AAH32865.1 | |||
BX640903.1 | CAE45949.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3893723 | Chr.1:73643288 - 74246497 on Build GRCh38 | Gain | FPGT TNNI3K LRRIQ3 FPGT-TNNI3K |
nsv1002115 | Chr.1:72379872 - 74933684 on Build GRCh38 | Gain | LOC105378803 LINC01360 CRYZ LOC339468 FPGT ERICH3 TNNI3K LRRIQ3 ERICH3-AS1 TYW3 FPGT-TNNI3K |
nsv522147 | Chr.1:73872847 - 74518151 on Build GRCh38 | Loss | LOC105378803 FPGT TNNI3K LRRIQ3 FPGT-TNNI3K |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |