Assay Details
Target Gene Details
Entrez Gene ID: | 2327 |
Gene Name: | flavin containing monooxygenase 2 |
Gene Aliases: |
FMO1B1 |
Location: |
Chr.1:171185208-171212683 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FMO2 | NM_001301347.1 | NP_001288276.1 | ||
NM_001460.4 | NP_001451.2 | |||
AK304385.1 | ||||
AK312877.1 | ||||
AL833218.1 | CAI46191.1 | |||
BC005894.1 | AAH05894.1 | |||
BI766311.1 | ||||
BT006979.1 | AAP35625.1 | |||
CA312067.1 | ||||
DB224311.1 | ||||
DB236882.1 | ||||
Y09267.1 | CAA70462.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv28731 | Chr.1:171046945 - 171757821 on Build GRCh38 | Gain+Loss | FMO1 PRRC2C FMO4 FMO2 MROH9 MIR1295B MIR1295A FMO3 VAMP4 FMO6P TOP1P1 MYOC |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |