Assay Details
Target Gene Details
Entrez Gene ID: | 4194 |
Gene Name: | MDM4, p53 regulator |
Gene Aliases: |
HDMX, MDMX, MRP1 |
Location: |
Chr.1:204516379-204558120 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MDM4 | NM_001204171.1 | NP_001191100.1 | ||
NM_001204172.1 | NP_001191101.1 | |||
NM_001278516.1 | NP_001265445.1 | |||
NM_001278517.1 | NP_001265446.1 | |||
NM_001278518.1 | NP_001265447.1 | |||
NM_001278519.1 | NP_001265448.1 | |||
NM_002393.4 | NP_002384.2 | |||
XM_017001311.1 | XP_016856800.1 | |||
XM_017001312.1 | XP_016856801.1 | |||
XM_017001313.1 | XP_016856802.1 | |||
AF007111.1 | AAB62928.1 | |||
AK223228.1 | BAD96948.1 | |||
AK303626.1 | ||||
AK304698.1 | ||||
AK310607.1 | ||||
AK310903.1 | ||||
AY923176.1 | AAY22054.1 | |||
AY949028.1 | AAX51227.1 | |||
BC067299.1 | AAH67299.1 | |||
BC105106.1 | AAI05107.1 | |||
BP247507.1 | ||||
BX640923.1 | CAE45961.1 | |||
CR749470.1 | CAH18300.1 | |||
KF435149.1 | ||||
KF435150.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |