Assay Details
Target Gene Details
Entrez Gene ID: | 255738 |
Gene Name: | proprotein convertase subtilisin/kexin type 9 |
Gene Aliases: |
FH3, HCHOLA3, LDLCQ1, NARC-1, NARC1, PC9 |
Location: |
Chr.1:55039476-55064853 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PCSK9 | NM_174936.3 | NP_777596.2 | ||
NR_110451.1 | ||||
AK075365.1 | BAC11572.1 | |||
AK122717.1 | ||||
AK293870.1 | ||||
AK297473.1 | ||||
EF692496.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv518638 | Chr.1:55044199 - 55060623 on Build GRCh38 | Loss |
![]() |
nsv527129 | Chr.1:55034756 - 55058311 on Build GRCh38 | Loss |
![]() |
nsv817614 | Chr.1:54580862 - 55735351 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)