Assay Details
Target Gene Details
Entrez Gene ID: | 128338 |
Gene Name: | DNA damage regulated autophagy modulator 2 |
Gene Aliases: |
CORD21, PRO180, TMEM77, WWFQ154 |
Location: |
Chr.1:111117332-111140216 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DRAM2 | NM_178454.4 | NP_848549.3 | ||
XM_005270469.2 | XP_005270526.1 | |||
XM_005270470.1 | XP_005270527.1 | |||
XM_006710361.1 | XP_006710424.1 | |||
XM_006710362.1 | XP_006710425.1 | |||
XM_011540707.1 | XP_011539009.1 | |||
XM_011540708.1 | XP_011539010.1 | |||
XM_017000320.1 | XP_016855809.1 | |||
XM_017000321.1 | XP_016855810.1 | |||
AK309557.1 | ||||
AL833098.1 | ||||
AY336747.1 | AAR02410.1 | |||
AY358492.1 | AAQ88856.1 | |||
BC047025.1 | AAH47025.3 | |||
BC091509.1 | AAH91509.1 | |||
DC339928.1 | ||||
EF710624.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv10639 | Chr.1:111126227 - 111244431 on Build GRCh38 | Gain | DENND2D CEPT1 DRAM2 CHI3L2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |