Assay Details
Target Gene Details
Entrez Gene ID: | 81627 |
Gene Name: | tRNA methyltransferase 1 like |
Gene Aliases: |
C1orf25, MST070, MSTP070, TRM1L, bG120K12.3 |
Location: |
Chr.1:185118085-185157098 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TRMT1L | NM_001202423.1 | NP_001189352.1 | ||
NM_030934.4 | NP_112196.3 | |||
XM_011510028.2 | XP_011508330.1 | |||
AF163261.1 | AAQ13590.1 | |||
AF288399.1 | AAG60618.1 | |||
AK074936.1 | BAC11302.1 | |||
AK074993.1 | BAC11341.1 | |||
AK098359.1 | ||||
AK131460.1 | BAD18605.1 | |||
AK226103.1 | ||||
AK302166.1 | ||||
AK312801.1 | ||||
BC045535.1 | AAH45535.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv548396 | Chr.1:185068972 - 185182246 on Build GRCh38 | Loss | SWT1 TRMT1L RNF2 |
nsv521613 | Chr.1:185102699 - 185202737 on Build GRCh38 | Loss | SWT1 TRMT1L |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |