Assay Details
Target Gene Details
Entrez Gene ID: | 5768 |
Gene Name: | quiescin sulfhydryl oxidase 1 |
Gene Aliases: |
Q6, QSCN6 |
Location: |
Chr.1:180154833-180198034 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
QSOX1 | NM_001004128.2 | NP_001004128.1 | ||
NM_002826.4 | NP_002817.2 | |||
AB209280.1 | BAD92517.1 | |||
AF361868.1 | AAM00263.1 | |||
AK290842.1 | ||||
AK290887.1 | ||||
AY358941.1 | AAQ89300.1 | |||
BC017692.1 | AAH17692.1 | |||
BC100023.1 | AAI00024.1 | |||
BX382719.2 | ||||
L42379.1 | AAA89173.1 | |||
U97276.2 | AAC09010.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv475n100 | Chr.1:180156531 - 180210921 on Build GRCh38 | Gain | QSOX1 FLJ23867 |
dgv476n100 | Chr.1:180157303 - 180231969 on Build GRCh38 | Gain | QSOX1 LHX4 FLJ23867 |
dgv474n100 | Chr.1:180152345 - 180273524 on Build GRCh38 | Gain | QSOX1 LHX4 LHX4-AS1 ACBD6 FLJ23867 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |