Assay Details
Target Gene Details
Entrez Gene ID: | 23566 |
Gene Name: | lysophosphatidic acid receptor 3 |
Gene Aliases: |
EDG7, Edg-7, GPCR, HOFNH30, LP-A3, LPA3, RP4-678I3 |
Location: |
Chr.1:84813403-84893213 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LPAR3 | NM_012152.2 | NP_036284.1 | ||
AF127138.1 | AAD56311.1 | |||
AF186380.1 | AAF00530.1 | |||
AF236117.1 | AAF91291.1 | |||
AK308283.1 | ||||
AY322547.1 | AAP84360.1 | |||
BC126268.1 | ||||
BC126270.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv830448 | Chr.1:84828411 - 85025427 on Build GRCh38 | Loss | MCOLN3 MCOLN2 LPAR3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |