Assay Details
Target Gene Details
Entrez Gene ID: | 9473 |
Gene Name: | thymocyte selection associated family member 2 |
Gene Aliases: |
C1orf38, ICB-1 |
Location: |
Chr.1:27872531-27886685 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
THEMIS2 | NM_001039477.2 | NP_001034566.1 | ||
NM_001105556.2 | NP_001099026.1 | |||
NM_001286113.1 | NP_001273042.1 | |||
NM_001286115.1 | NP_001273044.1 | |||
NM_004848.3 | NP_004839.2 | |||
XM_005246041.3 | XP_005246098.1 | |||
XM_006711050.1 | XP_006711113.1 | |||
XM_011542445.2 | XP_011540747.1 | |||
AK303090.1 | ||||
AK303141.1 | ||||
AL554503.3 | ||||
BC031655.1 | ||||
BC132692.1 | ||||
BC133049.1 | ||||
BG716895.1 | ||||
BM146655.1 | ||||
CR749321.1 | CAH18176.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv509069 | Chr.1:27817460 - 27905956 on Build GRCh38 | Insertion | SCARNA1 STX12 THEMIS2 PPP1R8 RPA2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |