Assay Details
Target Gene Details
Entrez Gene ID: | 148932 |
Gene Name: | MOB kinase activator 3C |
Gene Aliases: |
MOB1E, MOBKL2C |
Location: |
Chr.1:46607715-46616891 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MOB3C | NM_145279.4 | NP_660322.2 | ||
NM_201403.2 | NP_958805.1 | |||
XM_017000407.1 | XP_016855896.1 | |||
AJ580637.1 | CAE45269.1 | |||
AK090453.1 | ||||
AK091808.1 | BAC03752.1 | |||
BC121169.2 | ||||
BG717827.1 | ||||
BI601393.1 | ||||
CF125162.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1009193 | Chr.1:46474327 - 46619995 on Build GRCh38 | Gain | MKNK1-AS1 KNCN MKNK1 DMBX1 MOB3C |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |