Assay Details
Target Gene Details
Entrez Gene ID: | 286826 |
Gene Name: | lin-9 DREAM MuvB core complex component |
Gene Aliases: |
BARA, BARPsv, Lin-9, TGS, TGS1, TGS2 |
Location: |
Chr.1:226231149-226310082 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LIN9 | NM_001270409.1 | NP_001257338.1 | ||
NM_001270410.1 | NP_001257339.1 | |||
NM_173083.3 | NP_775106.2 | |||
XM_005273102.3 | XP_005273159.1 | |||
XM_006711766.2 | XP_006711829.2 | |||
XM_006711767.3 | XP_006711830.1 | |||
XM_017001084.1 | XP_016856573.1 | |||
AF190323.1 | AAQ13710.1 | |||
AF190324.1 | AAQ13711.1 | |||
AK125690.1 | ||||
AK126177.1 | BAC86475.1 | |||
AK308823.1 | ||||
AY786184.1 | AAV41873.1 | |||
BC043444.2 | AAH43444.1 | |||
BC045625.1 | AAH45625.1 | |||
BC065302.1 | AAH65302.1 | |||
BX537869.1 | CAD97871.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv517107 | Chr.1:226187566 - 226320944 on Build GRCh38 | Gain+Loss | MIXL1 LIN9 LOC101927247 |
esv3891148 | Chr.1:226235871 - 226306814 on Build GRCh38 | Loss | LIN9 |
nsv832759 | Chr.1:226116059 - 226316469 on Build GRCh38 | Loss | MIXL1 ACBD3 LIN9 LOC101927247 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |