Assay Details
Target Gene Details
Entrez Gene ID: | 23352 |
Gene Name: | ubiquitin protein ligase E3 component n-recognin 4 |
Gene Aliases: |
RBAF600, ZUBR1, p600 |
Location: |
Chr.1:19074506-19210334 on Build GRCh38 |
Assay Gene Location: | Within Intron 104 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834347 | Chr.1:18916277 - 19142428 on Build GRCh38 | Gain | IFFO2 UBR4 |
nsv508980 | Chr.1:19042348 - 19097757 on Build GRCh38 | Insertion | UBR4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |