Assay Details
Target Gene Details
Entrez Gene ID: | 1718 |
Gene Name: | 24-dehydrocholesterol reductase |
Gene Aliases: |
DCE, Nbla03646, SELADIN1, seladin-1 |
Location: |
Chr.1:54849627-54887248 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DHCR24 | NM_014762.3 | NP_055577.1 | ||
AB073391.1 | BAE45721.1 | |||
AF261758.1 | AAG17288.1 | |||
AK298414.1 | ||||
AK302774.1 | ||||
AK304302.1 | ||||
AK304421.1 | ||||
AK304437.1 | ||||
AK316419.1 | ||||
AK316427.1 | ||||
BC004375.1 | AAH04375.1 | |||
BC011669.2 | AAH11669.1 | |||
D13643.3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv546266 | Chr.1:54404143 - 54887697 on Build GRCh38 | Gain | MROH7 FAM151A TTC22 SSBP3 PARS2 MROH7-TTC4 ACOT11 DHCR24 LEXM TTC4 |
nsv817614 | Chr.1:54580862 - 55735351 on Build GRCh38 | Gain | BSND TMEM61 USP24 PCSK9 DHCR24 LOC100507634 MIR4422 MROH7 FAM151A TTC22 PARS2 MROH7-TTC4 ACOT11 LEXM TTC4 |
nsv829915 | Chr.1:54828940 - 54992690 on Build GRCh38 | Gain | TMEM61 DHCR24 LEXM |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |