Assay Details
Target Gene Details
Entrez Gene ID: | 7412 |
Gene Name: | vascular cell adhesion molecule 1 |
Gene Aliases: |
CD106, INCAM-100 |
Location: |
Chr.1:100719640-100739045 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
VCAM1 | NM_001078.3 | NP_001069.1 | ||
NM_001199834.1 | NP_001186763.1 | |||
NM_080682.2 | NP_542413.1 | |||
AK223266.1 | BAD96986.1 | |||
AK291732.1 | ||||
AK296692.1 | ||||
BC017276.2 | AAH17276.3 | |||
BC068490.1 | AAH68490.2 | |||
BC085003.1 | AAH85003.1 | |||
M30257.1 | AAA51917.1 | |||
M60335.1 | AAA61269.1 | |||
X53051.1 | CAA37218.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv522064 | Chr.1:100715786 - 101074310 on Build GRCh38 | Gain | LOC102606465 SLC30A7 VCAM1 DPH5 EXTL2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |