Assay Details
Target Gene Details
Entrez Gene ID: | 9372 |
Gene Name: | zinc finger FYVE-type containing 9 |
Gene Aliases: |
MADHIP, NSP, PPP1R173, SARA, SMADIP |
Location: |
Chr.1:52142001-52348664 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ZFYVE9 | NM_004799.3 | NP_004790.2 | ||
NM_007324.3 | NP_015563.2 | |||
XM_017002845.1 | XP_016858334.1 | |||
AF130419.1 | AAD31694.1 | |||
AF130420.1 | AAD31695.1 | |||
DR005493.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv855 | Chr.1:52155519 - 52187874 on Build GRCh38 | Insertion | ZFYVE9 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |